Learn More
Waters Corp Waters XBridge Premier Oligonucleotide, BEH C18, 130 Å, 2.5 μm, 4.6 x 100 mm
Catalog No. 502224319 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XBridge Premier Oligonucleotide column is designed for advanced bioseparations, specifically tailored for oligonucleotide analysis. It features BEH C18 particle technology for robust performance.
  • Offers outstanding peak capacity, superior peak shape, and extended lifetime when used with HFIP, HAA, and TEA.
  • Provides two pore sizes (130 Å and 300 Å) to achieve different separation selectivities.
  • Available with optional VanGuard FIT Column technology.
Catalog No. 50-222-4319 Supplier Waters Corp Supplier No. 186009902
This XBridge Premier Oligonucleotide column is designed for advanced bioseparations, specifically tailored for oligonucleotide analysis. It features BEH C18 particle technology for robust performance.
  • Offers outstanding peak capacity, superior peak shape, and extended lifetime when used with HFIP, HAA, and TEA.
  • Provides two pore sizes (130 Å and 300 Å) to achieve different separation selectivities.
  • Available with optional VanGuard FIT Column technology.