Learn More
Waters Corp Waters Column, Oligonucleotide BEH C18, 2.5 μm, 2.1 x 50 mm
Catalog No. 502224255 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Premier Oligonucleotide Columns separate detritylated synthetic oligonucleotides via ion-pair, reversed-phase chromatography. Featuring Ethylene Bridged Hybrid (BEH) particles and MaxPeak High Performance Surface technology, they enhance reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Designed for ion-pair, reversed-phase chromatography of detritylated synthetic oligonucleotides, with QC testing for repeatable performance.
  • MaxPeak Premier Class improves reproducibility and peak shape for metal-sensitive analytes.
  • Utilizes Ethylene Bridged Hybrid (BEH) particles and MaxPeak High Performance Surface technology.
  • Extendable lifetime with VanGuard FIT technology; offers high pH stability in UHPLC.
  • Specifications: 2.5 μm particle size, 130 Å pore size, 2.1 mm ID, 50 mm length.
  • Max pressure: 18000 psi (1240 Bar); pH range: 1 to 12.
Catalog No. 50-222-4255 Supplier Waters Corp Supplier No. 186009836
These Premier Oligonucleotide Columns separate detritylated synthetic oligonucleotides via ion-pair, reversed-phase chromatography. Featuring Ethylene Bridged Hybrid (BEH) particles and MaxPeak High Performance Surface technology, they enhance reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Designed for ion-pair, reversed-phase chromatography of detritylated synthetic oligonucleotides, with QC testing for repeatable performance.
  • MaxPeak Premier Class improves reproducibility and peak shape for metal-sensitive analytes.
  • Utilizes Ethylene Bridged Hybrid (BEH) particles and MaxPeak High Performance Surface technology.
  • Extendable lifetime with VanGuard FIT technology; offers high pH stability in UHPLC.
  • Specifications: 2.5 μm particle size, 130 Å pore size, 2.1 mm ID, 50 mm length.
  • Max pressure: 18000 psi (1240 Bar); pH range: 1 to 12.