Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XBridge VanGuard FIT Cartridge, C18, 2.5 μm, 2.1 x 5 mm

Catalog No. 502391728 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

An XBridge Premier Peptide BEH C18 VanGuard FIT Cartridge, 2.5 μm, 2.1 X 5mm is a replacement cartridge designed for XBridge Premier Peptide BEH C18 Columns with the Premier VanGuard FIT column design. It is recommended for replacement when there is evidence of high analytical column backpressure or a reduction in desired component resolution.

  • C18 chemistry for reversed phase separation.
  • Hybrid particle substrate with spherical particle shape.
  • pH range of 1 to 12 pH.
  • Temperature limit of 80°C.
  • Maximum pressure of 18000 psi (1240 bar).
  • Endcapped design with dC18 bonding technology.
  • Low silanol activity.
  • 2.5 μm particle size.
  • 130 Å pore size.
  • Parker-style endfitting.
  • Peptide QC tested.
  • UPLC system compatible.
  • LC technique for peptide applications.
  • Supplied as 3 units per package.

Catalog No. 50-239-1728 Supplier Waters Corp Supplier No. 186010706
May include imposed supplier surcharges.
Add to Cart
Add to Cart

An XBridge Premier Peptide BEH C18 VanGuard FIT Cartridge, 2.5 μm, 2.1 X 5mm is a replacement cartridge designed for XBridge Premier Peptide BEH C18 Columns with the Premier VanGuard FIT column design. It is recommended for replacement when there is evidence of high analytical column backpressure or a reduction in desired component resolution.

  • C18 chemistry for reversed phase separation.
  • Hybrid particle substrate with spherical particle shape.
  • pH range of 1 to 12 pH.
  • Temperature limit of 80°C.
  • Maximum pressure of 18000 psi (1240 bar).
  • Endcapped design with dC18 bonding technology.
  • Low silanol activity.
  • 2.5 μm particle size.
  • 130 Å pore size.
  • Parker-style endfitting.
  • Peptide QC tested.
  • UPLC system compatible.
  • LC technique for peptide applications.
  • Supplied as 3 units per package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.