Learn More
Waters Corp Waters Corporation XBridge Premier Peptide BEH C18 Column, 300 Å, 2.5 μm, 4.6 mm x 150 mm
Catalog No. 502224314 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Premier Peptide BEH C18 Column is optimized and QC tested for peptide separations. Its wide-pore (300Å) trifunctionally bonded BEH particle offers a wide usable pH range, superior low pH stability, and ultra-low column bleed, making it suitable for large peptide analysis in proteomics, protein characterization, and peptide synthesis. It is part of the MaxPeak Premier Class of Columns, designed to improve reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Specially optimized and QC tested with cytochrome c digest standards for peptide separations.
  • Wide-pore (300Å) trifunctionally bonded BEH particle.
  • Offers the widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed.
  • Delivers chromatographic performance while increasing reproducibility, peak shape, and recovery for metal-sensitive analytes.
Catalog No. 50-222-4314 Supplier Waters Corp Supplier No. 186009897
The XBridge Premier Peptide BEH C18 Column is optimized and QC tested for peptide separations. Its wide-pore (300Å) trifunctionally bonded BEH particle offers a wide usable pH range, superior low pH stability, and ultra-low column bleed, making it suitable for large peptide analysis in proteomics, protein characterization, and peptide synthesis. It is part of the MaxPeak Premier Class of Columns, designed to improve reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Specially optimized and QC tested with cytochrome c digest standards for peptide separations.
  • Wide-pore (300Å) trifunctionally bonded BEH particle.
  • Offers the widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed.
  • Delivers chromatographic performance while increasing reproducibility, peak shape, and recovery for metal-sensitive analytes.