Learn More
Waters Corp XBridge Column, C4, 300 Å, 2.5 μm, 4.6 x 50 mm
Catalog No. 502390706 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Optimized for protein separation based on size, hydrophobicity, and isoelectric point.
  • C4 ligand minimizes protein carryover, increasing recovery and peak capacity.
  • MaxPeak Premier Class columns enhance reproducibility and peak shape for metal-sensitive analytes.
  • Includes electronic eConnect tag for automatic identification and usage tracking.
  • eConnect columns realize full advantages with Alliance iS/Bio System running Empower software.
  • QC tested using MassPREP Protein Mix standard.
  • Suggested for low-level intact protein/subunit analysis and phosphorylated proteins.
Catalog No. 50-239-0706 Supplier Waters Corp Supplier No. 186010332
  • Optimized for protein separation based on size, hydrophobicity, and isoelectric point.
  • C4 ligand minimizes protein carryover, increasing recovery and peak capacity.
  • MaxPeak Premier Class columns enhance reproducibility and peak shape for metal-sensitive analytes.
  • Includes electronic eConnect tag for automatic identification and usage tracking.
  • eConnect columns realize full advantages with Alliance iS/Bio System running Empower software.
  • QC tested using MassPREP Protein Mix standard.
  • Suggested for low-level intact protein/subunit analysis and phosphorylated proteins.