Learn More
Waters Corp XBridge Premier Protein BEH C4 Column, 300 Å, 2.5 μm, 2.1 x 100 mm
Catalog No. 502392997 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Premier Protein BEH C4 300 Å columns are QC tested with the MassPREP Protein Mix standard and are optimized to separate proteins based on size, hydrophobicity, and isoelectric point.
  • C4 ligand offers less retention compared to traditional C18 phases, minimizing protein carryover, increasing protein recovery, and improving peak capacity.
  • Part of the MaxPeak Premier Class of Columns, providing chromatographic performance with increased reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Includes a vial of MassPREP Protein Mix Standard at no additional cost.
Catalog No. 50-239-2997 Supplier Waters Corp Supplier No. 176005111
The XBridge Premier Protein BEH C4 300 Å columns are QC tested with the MassPREP Protein Mix standard and are optimized to separate proteins based on size, hydrophobicity, and isoelectric point.
  • C4 ligand offers less retention compared to traditional C18 phases, minimizing protein carryover, increasing protein recovery, and improving peak capacity.
  • Part of the MaxPeak Premier Class of Columns, providing chromatographic performance with increased reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Includes a vial of MassPREP Protein Mix Standard at no additional cost.