Learn More
Waters Corp Column, C4, 300 Å, 2.5 μm, 4.6 x 150 mm
Catalog No. 502393028 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is optimized for separating proteins by size, hydrophobicity, and isoelectric point. Its C4 ligand offers less retention than C18 phases, minimizing protein carryover and improving recovery and peak capacity. As a MaxPeak Premier Class Column, it delivers consistent chromatographic performance, enhanced reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Separates proteins of various sizes, hydrophobicities, and isoelectric points.
  • QC tested with MassPREP Protein Mix standard.
  • Provides consistent chromatography with enhanced reproducibility and peak shape.
  • Improves recovery for metal-sensitive analytes; minimizes protein carryover.
  • Tolerates extreme pH and temperature.
  • Minimizes secondary interactions.
  • Enhances sensitivity for phosphorylated proteins and low-level intact/subunit mAb analyses.
Catalog No. 50-239-3028 Supplier Waters Corp Supplier No. 176005115
This column is optimized for separating proteins by size, hydrophobicity, and isoelectric point. Its C4 ligand offers less retention than C18 phases, minimizing protein carryover and improving recovery and peak capacity. As a MaxPeak Premier Class Column, it delivers consistent chromatographic performance, enhanced reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Separates proteins of various sizes, hydrophobicities, and isoelectric points.
  • QC tested with MassPREP Protein Mix standard.
  • Provides consistent chromatography with enhanced reproducibility and peak shape.
  • Improves recovery for metal-sensitive analytes; minimizes protein carryover.
  • Tolerates extreme pH and temperature.
  • Minimizes secondary interactions.
  • Enhances sensitivity for phosphorylated proteins and low-level intact/subunit mAb analyses.