Learn More
Waters Corp Columns, Protein BEH C4, 300 Å Pore, 2.5 μm Particle, 2.1 x 100 mm
Catalog No. 502391168 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Premier Protein BEH C4 300 Å 2.5 μm, 2.1 x 100 mm columns are designed for the separation of proteins based on size, hydrophobicity, and isoelectric point. They feature a C4 ligand for reduced retention, minimized protein carryover, increased recovery, and improved peak capacity, especially beneficial for MS sensitivity at low levels. These columns are part of the MaxPeak Premier Class, offering enhanced reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • 2.1 mm ID columns recommended for low dispersion (UPLC or UHPLC) systems, especially for LC-MS methods.
  • QC tested with the MassPREP Protein Mix standard.
  • Suggested for low-level intact protein and subunit analysis and phosphorylated proteins.
Catalog No. 50-239-1168 Supplier Waters Corp Supplier No. 186010330
Add to Cart
Add to Cart
The XBridge Premier Protein BEH C4 300 Å 2.5 μm, 2.1 x 100 mm columns are designed for the separation of proteins based on size, hydrophobicity, and isoelectric point. They feature a C4 ligand for reduced retention, minimized protein carryover, increased recovery, and improved peak capacity, especially beneficial for MS sensitivity at low levels. These columns are part of the MaxPeak Premier Class, offering enhanced reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • 2.1 mm ID columns recommended for low dispersion (UPLC or UHPLC) systems, especially for LC-MS methods.
  • QC tested with the MassPREP Protein Mix standard.
  • Suggested for low-level intact protein and subunit analysis and phosphorylated proteins.