Learn More
Waters Corp Column, BEH C4, 300 Å/2.5 μm, 2.1 x 50 mm
Catalog No. 502392749 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge Premier Protein BEH C4 300 Å 2.5 μm, 2.1 x 50 mm columns are optimized for protein separation by size, hydrophobicity, and isoelectric point. Their C4 ligand reduces protein carryover, improves recovery, and enhances peak capacity. These MaxPeak Premier Class columns also boost MS sensitivity and improve reproducibility for metal-sensitive analytes.
  • Optimized for protein separation.
  • C4 ligand reduces protein carryover and improves recovery.
  • Enhances peak capacity and MS sensitivity.
  • MaxPeak Premier Class for improved reproducibility with metal-sensitive analytes.
  • QC tested with MassPREP Protein Mix standard.
  • All column documents available online.
Catalog No. 50-239-2749 Supplier Waters Corp Supplier No. 186010329
XBridge Premier Protein BEH C4 300 Å 2.5 μm, 2.1 x 50 mm columns are optimized for protein separation by size, hydrophobicity, and isoelectric point. Their C4 ligand reduces protein carryover, improves recovery, and enhances peak capacity. These MaxPeak Premier Class columns also boost MS sensitivity and improve reproducibility for metal-sensitive analytes.
  • Optimized for protein separation.
  • C4 ligand reduces protein carryover and improves recovery.
  • Enhances peak capacity and MS sensitivity.
  • MaxPeak Premier Class for improved reproducibility with metal-sensitive analytes.
  • QC tested with MassPREP Protein Mix standard.
  • All column documents available online.