Learn More
Waters Corp Column, XBridge Premier Protein BEH C4, 300 Å, 2.5 μm
Supplier:  Waters Corp 176005112
Encompass_Preferred
The XBridge Premier Protein BEH C4 300 Å columns are QC tested with MassPREP Protein Mix standard, optimized for protein separation by size, hydrophobicity, and isoelectric point. The C4 ligand provides less retention than C18 phases, minimizing protein carryover, increasing recovery, and improving peak capacity. As MaxPeak Premier Class Columns, they enhance reproducibility, peak shape, and recovery for metal-sensitive analytes. A MassPREP Protein Mix Standard vial is included.
  • Optimized for protein separation by size, hydrophobicity, and isoelectric point.
  • QC tested with MassPREP Protein Mix standard.
  • C4 ligand provides reduced retention.
  • Minimizes protein carryover and boosts protein recovery and peak capacity.
  • Ensures enhanced reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Includes a MassPREP Protein Mix Standard vial.
Catalog No. 50-239-2886