Learn More
Waters Corp XBridge Premier Protein SEC Column, 250Å, 2.5 μm, 4.6 x 150 mm
Supplier:  Waters Corp 176004790
Encompass_Preferred
XBridge Premier Protein SEC 250Å, 2.5 μm columns provide reliable protein size variant separations (10,000 to 650,000 Daltons). Combining MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies, these columns reduce secondary ionic or hydrophobic interactions, minimizing method development.
  • Reliable protein size variant separations (10,000 to 650,000 Daltons).
  • Utilizes MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies.
  • Minimizes secondary ionic or hydrophobic interactions.
  • Reduces method development with a “generic” or “platform-type” method.
  • Includes one vial of mAb size variant standard.
  • Includes the MaxPeak Premier Protein SEC 250Å, 2.5 μm, 4.6 x 30 mm guard.
Catalog No. 50-239-2337