Learn More
Waters Corp XBridge Premier SEC Column, 250Å 2.5 μm, 4.6 × 300 mm
Catalog No. 502392481 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge Premier Protein SEC 250Å, 2.5 μm columns are designed for reliable protein size variant separations from 10,000 to 650,000 Daltons. They leverage MaxPeak Premier High Performance Surfaces and BEH-PEO particle technologies to minimize undesirable secondary ionic or hydrophobic interactions, reducing the need for extensive method development and supporting generic methods.
  • Designed for reliable protein size variant separations (10,000 to 650,000 Daltons).
  • Utilizes MaxPeak Premier high performance surfaces and BEH-PEO particle technologies.
  • Minimizes secondary ionic and hydrophobic interactions.
  • Reduces extensive method development and supports generic methods.
  • Provides 30% improved resolution with narrow peak widths.
  • Offers exceptional bioinertness and improved sensitivity.
  • Ensures reliable quantification of high molecular weight species.
Catalog No. 50-239-2481 Supplier Waters Corp Supplier No. 176004780
XBridge Premier Protein SEC 250Å, 2.5 μm columns are designed for reliable protein size variant separations from 10,000 to 650,000 Daltons. They leverage MaxPeak Premier High Performance Surfaces and BEH-PEO particle technologies to minimize undesirable secondary ionic or hydrophobic interactions, reducing the need for extensive method development and supporting generic methods.
  • Designed for reliable protein size variant separations (10,000 to 650,000 Daltons).
  • Utilizes MaxPeak Premier high performance surfaces and BEH-PEO particle technologies.
  • Minimizes secondary ionic and hydrophobic interactions.
  • Reduces extensive method development and supports generic methods.
  • Provides 30% improved resolution with narrow peak widths.
  • Offers exceptional bioinertness and improved sensitivity.
  • Ensures reliable quantification of high molecular weight species.