Learn More
Waters Corp XBridge Premier Protein SEC Column, 250 Å, 2.5 μm, 4.6 mm x 300 mm
Catalog No. 502390540 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These SEC columns enable reliable protein size variant separations for proteins from 10,000 to 650,000 Daltons. Combining MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies, these advancements reduce undesirable secondary ionic or hydrophobic interactions, minimizing method development.
  • Assists in achieving reliable protein size variant separations for proteins ranging from 10,000 to approximately 650,000 Daltons.
  • Utilizes MaxPeak Premier High Performance Surfaces.
  • Incorporates innovative BEH-PEO Particle technologies.
  • Designed to reduce undesirable secondary ionic or hydrophobic interactions.
  • Minimizes extensive method development work.
  • Allows for “generic” or “platform-type” method for various samples.
  • Each batch is specifically QC tested to ensure consistent performance for challenging size-based protein separations.
Catalog No. 50-239-0540 Supplier Waters Corp Supplier No. 186009960
These SEC columns enable reliable protein size variant separations for proteins from 10,000 to 650,000 Daltons. Combining MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies, these advancements reduce undesirable secondary ionic or hydrophobic interactions, minimizing method development.
  • Assists in achieving reliable protein size variant separations for proteins ranging from 10,000 to approximately 650,000 Daltons.
  • Utilizes MaxPeak Premier High Performance Surfaces.
  • Incorporates innovative BEH-PEO Particle technologies.
  • Designed to reduce undesirable secondary ionic or hydrophobic interactions.
  • Minimizes extensive method development work.
  • Allows for “generic” or “platform-type” method for various samples.
  • Each batch is specifically QC tested to ensure consistent performance for challenging size-based protein separations.