Learn More
Waters Corp XBridge Premier Protein SEC Column, 250Å, 2.5 μm, 7.8 x 300 mm
Catalog No. 502390743 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge Premier Protein SEC 250Å, 2.5 μm Columns provide reliable protein size variant separations (10,000 to approximately 650,000 Daltons). Combining MaxPeak Premier High Performance Surfaces and novel BEH-PEO Particle technologies, these columns minimize undesired secondary ionic or hydrophobic interactions. This helps reduce method development costs by enabling a “generic” or “platform-type” method for various samples.
  • cGMP, ISO 9001 certified manufacturing with stringent QC.
  • Provides method flexibility and robustness across buffer ranges.
  • Minimizes hydrophobic secondary interactions, ideal for LC-MS applications.
  • Enhances chromatography with ammonium acetate mobile phases, improving sample recoveries.
  • Features high coverage bondings.
  • Includes QR code on column label and optional electronic column tags.
Catalog No. 50-239-0743 Supplier Waters Corp Supplier No. 176005070
Add to Cart
Add to Cart
XBridge Premier Protein SEC 250Å, 2.5 μm Columns provide reliable protein size variant separations (10,000 to approximately 650,000 Daltons). Combining MaxPeak Premier High Performance Surfaces and novel BEH-PEO Particle technologies, these columns minimize undesired secondary ionic or hydrophobic interactions. This helps reduce method development costs by enabling a “generic” or “platform-type” method for various samples.
  • cGMP, ISO 9001 certified manufacturing with stringent QC.
  • Provides method flexibility and robustness across buffer ranges.
  • Minimizes hydrophobic secondary interactions, ideal for LC-MS applications.
  • Enhances chromatography with ammonium acetate mobile phases, improving sample recoveries.
  • Features high coverage bondings.
  • Includes QR code on column label and optional electronic column tags.