Learn More
Waters Corp XBridge Premier Protein SEC Column, 250 Å, 2.5 μm, 4.6 x 150 mm
Catalog No. 502391650 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge Premier Protein SEC 250Å, 2.5 μm Columns provide reliable protein size variant separations for molecular weights from 10,000 to approximately 650,000 Daltons. Utilizing MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies, these columns minimize unwanted secondary ionic or hydrophobic interactions, reducing extensive method development and allowing for a “generic” or “platform-type” method for various samples.
  • Reliable protein size variant separations for molecular weights 10,000 to 650,000 Daltons.
  • Minimizes unwanted secondary ionic or hydrophobic interactions.
  • Reduces need for extensive method development.
  • Enables “generic” or “platform-type” methods for diverse samples.
Catalog No. 50-239-1650 Supplier Waters Corp Supplier No. 176004779
Add to Cart
Add to Cart
XBridge Premier Protein SEC 250Å, 2.5 μm Columns provide reliable protein size variant separations for molecular weights from 10,000 to approximately 650,000 Daltons. Utilizing MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies, these columns minimize unwanted secondary ionic or hydrophobic interactions, reducing extensive method development and allowing for a “generic” or “platform-type” method for various samples.
  • Reliable protein size variant separations for molecular weights 10,000 to 650,000 Daltons.
  • Minimizes unwanted secondary ionic or hydrophobic interactions.
  • Reduces need for extensive method development.
  • Enables “generic” or “platform-type” methods for diverse samples.