Learn More
Waters Corp XBridge Premier Protein SEC Column, 250Å, 2.5 μm, 7.8 x 150 mm
Supplier:  Waters Corp 176005069
Encompass_Preferred
These SEC 250Å, 2.5 μm columns achieve reliable protein size variant separations (10,000 to 650,000 Daltons) by combining MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies. This design minimizes undesirable secondary ionic or hydrophobic interactions, streamlining method development for “generic” or “platform-type” samples. Each column includes one vial of mAb Size Variant Standard.
  • Reliable protein size variant separations (10,000 to approximately 650,000 Daltons).
  • Minimized undesirable secondary ionic or hydrophobic interactions.
  • Streamlined method development with “generic” or “platform-type” methods.
  • Includes one vial of mAb Size Variant Standard.
Catalog No. 50-239-1923