Learn More
Waters Corp Prep Guard Cartridge, Amide, 5 μm, 10 x 10 mm
Catalog No. 50466350 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard cartridge is designed for high-performance liquid chromatography, effectively retaining extremely polar compounds such as carbohydrates and sugars. It features a chemically stable, trifunctionally bonded amide phase, stable from pH 2 to 11.
  • Retains extremely polar compounds including carbohydrates and sugars.
  • Features a chemically stable amide phase, stable from pH 2 to 11.
  • Constructed with Ethylene Bridged Hybrid (BEH) for mechanical strength and chemical stability.
  • BEH particle technology ensures maximum column performance and longer lifetimes.
  • Hybrid particle technology offers improved reproducibility, peak shape, and efficiency.
  • Allows for seamless transfer from analytical to preparative separations.
Catalog No. 50-466-350 Supplier Waters Corp Supplier No. 186006597
This guard cartridge is designed for high-performance liquid chromatography, effectively retaining extremely polar compounds such as carbohydrates and sugars. It features a chemically stable, trifunctionally bonded amide phase, stable from pH 2 to 11.
  • Retains extremely polar compounds including carbohydrates and sugars.
  • Features a chemically stable amide phase, stable from pH 2 to 11.
  • Constructed with Ethylene Bridged Hybrid (BEH) for mechanical strength and chemical stability.
  • BEH particle technology ensures maximum column performance and longer lifetimes.
  • Hybrid particle technology offers improved reproducibility, peak shape, and efficiency.
  • Allows for seamless transfer from analytical to preparative separations.