Learn More
Waters Corp Waters Corporation Prep Guard Cartridge, 130Å, 5 μm, 19 mm X 10 mm
Supplier:  Waters Corp 186006598
Encompass_Preferred
The XBridge BEH Amide Prep Guard Cartridge is designed to safeguard liquid chromatography (LC) systems from particulate matter. It features a chemically stable, trifunctionally bonded amide phase, ideal for retaining extremely polar compounds like carbohydrates and sugars, stable from pH 2 to 11. With a 130 Å pore size and 5 μm particle size, it ensures efficient particle removal and a long column lifetime for diverse applications.
  • Amine-bonded for separating polar compounds
  • Chemically stable amide phase, pH 2 to 11
  • 130 Å pore size and 5 μm particle size
  • Safeguards LC systems against particulate matter
  • Easy installation and use
  • Hydrophilic Interaction (HILIC) separation mode
  • Hybrid particle substrate
  • Max pressure of 6000 psi (415 Bar)
  • 19 mm inner diameter, 10 mm length
Catalog No. 50-466-351