Learn More
Waters Corp XBridge BEH Amide OBD Column, 130Å, 5 μm, 19 x 100 mm
Catalog No. 50466357 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These preparative columns are designed for the retention of extremely polar compounds, including carbohydrates and sugars. They feature a chemically stable, trifunctionally bonded amide phase, which ensures stability across a pH range of 2 to 11, enabling the separation of polar analytes with diverse polarity, structural characteristics, and pKa values.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Chemically stable, trifunctionally bonded amide phase.
  • Stable from pH 2 to 11.
  • Enables separation of polar analytes spanning a wide range of polarity, structural moiety, and pKa.
Catalog No. 50-466-357 Supplier Waters Corp Supplier No. 186006604
Add to Cart
Add to Cart
These preparative columns are designed for the retention of extremely polar compounds, including carbohydrates and sugars. They feature a chemically stable, trifunctionally bonded amide phase, which ensures stability across a pH range of 2 to 11, enabling the separation of polar analytes with diverse polarity, structural characteristics, and pKa values.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Chemically stable, trifunctionally bonded amide phase.
  • Stable from pH 2 to 11.
  • Enables separation of polar analytes spanning a wide range of polarity, structural moiety, and pKa.