Learn More
Waters Corp Waters XBridge BEH Amide OBD Prep Column, 5 μm, 19 × 150 mm
Catalog No. 50466358 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH Amide OBD Prep Column features 5 μm particles and dimensions of 19 × 150 mm, designed for the separation of polar analytes.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Chemically stable amide phase, stable from pH 2 to 11.
  • Enables separation of polar analytes across a wide range of polarity and pKa.
  • Boosts efficiency for lab-scale separations using optimum bed density (OBD) technology.
  • Scales directly from UPLC or HPLC screening to lab-scale purification.
  • Patented packing ensures uniform density profiles and repeatable column performance.
  • Exceptional durability with minimal retention loss and peak shape deterioration.
  • Withstands pressures up to 6,000 psi (415 Bar).
Catalog No. 50-466-358 Supplier Waters Corp Supplier No. 186006605
The XBridge BEH Amide OBD Prep Column features 5 μm particles and dimensions of 19 × 150 mm, designed for the separation of polar analytes.
  • Retains extremely polar compounds, including carbohydrates and sugars.
  • Chemically stable amide phase, stable from pH 2 to 11.
  • Enables separation of polar analytes across a wide range of polarity and pKa.
  • Boosts efficiency for lab-scale separations using optimum bed density (OBD) technology.
  • Scales directly from UPLC or HPLC screening to lab-scale purification.
  • Patented packing ensures uniform density profiles and repeatable column performance.
  • Exceptional durability with minimal retention loss and peak shape deterioration.
  • Withstands pressures up to 6,000 psi (415 Bar).