Learn More
Waters Corp Waters Corporation OBD Prep Column, Amide, 5 μm, 30 mm x 100 mm
Catalog No. 50466362 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
BEH Amide columns are designed to retain extremely polar compounds, including carbohydrates and sugars. They feature a chemically stable, trifunctionally bonded amide phase that maintains stability across a pH range of 2 to 11. This enables the separation of diverse polar analytes with varying polarity, structural characteristics, and pKa values.
  • Amide chemistry for retaining extremely polar compounds
  • Stable across a pH range of 2 to 11
  • Temperature limits up to 90 °C
  • Particle size of 5 μm
  • Pore size of 130 Å
  • Maximum pressure of 6000 psi (415 Bar)
  • Suitable for LC/MS and LC techniques
  • Provides alternate selectivity compared to straight-chain C18 for polar basic/polar acidic molecules
Catalog No. 50-466-362 Supplier Waters Corp Supplier No. 186006609
Add to Cart
Add to Cart
Request a Quote
BEH Amide columns are designed to retain extremely polar compounds, including carbohydrates and sugars. They feature a chemically stable, trifunctionally bonded amide phase that maintains stability across a pH range of 2 to 11. This enables the separation of diverse polar analytes with varying polarity, structural characteristics, and pKa values.
  • Amide chemistry for retaining extremely polar compounds
  • Stable across a pH range of 2 to 11
  • Temperature limits up to 90 °C
  • Particle size of 5 μm
  • Pore size of 130 Å
  • Maximum pressure of 6000 psi (415 Bar)
  • Suitable for LC/MS and LC techniques
  • Provides alternate selectivity compared to straight-chain C18 for polar basic/polar acidic molecules