Learn More
Waters Corp XBridge BEH C18 Prep Guard Cartridge, 130Å, 5 μm, 19 mm × 10 mm
Catalog No. 50781431 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Prep Guard Cartridge protects preparative columns by capturing particulates and strongly retained compounds. It features a trifunctionally bonded C18 ligand, making it suitable for most HPLC separations. It offers a wide usable pH range (1–12), superior low pH stability, and ultra–low column bleed, making it ideal for a broad range of hydrophobic compounds and high–throughput environments.
  • Protects preparative columns by capturing particulates and strongly retained compounds.
  • Suitable for an entire range of mobile phase pH (1–12).
  • Provides a universal column for most HPLC separations.
  • Offers superior low pH stability.
  • Ensures ultra–low column bleed.
  • Well–suited for a broad range of hydrophobic compounds.
  • Designed to withstand high–throughput environments, offering robustness and durability.
Catalog No. 50-781-431 Supplier Waters Corp Supplier No. 186002975
This Prep Guard Cartridge protects preparative columns by capturing particulates and strongly retained compounds. It features a trifunctionally bonded C18 ligand, making it suitable for most HPLC separations. It offers a wide usable pH range (1–12), superior low pH stability, and ultra–low column bleed, making it ideal for a broad range of hydrophobic compounds and high–throughput environments.
  • Protects preparative columns by capturing particulates and strongly retained compounds.
  • Suitable for an entire range of mobile phase pH (1–12).
  • Provides a universal column for most HPLC separations.
  • Offers superior low pH stability.
  • Ensures ultra–low column bleed.
  • Well–suited for a broad range of hydrophobic compounds.
  • Designed to withstand high–throughput environments, offering robustness and durability.