Learn More
Waters Corp Waters Preparative Column, XBridge BEH C18, 10 μm, 19 x 150 mm
Catalog No. 50782053 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 sorbent facilitates HPLC method development, usable across a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand provides universal HPLC separations with superior low pH stability and ultra-low column bleed. The Optimum Bed Density (OBD) design enables faster, more effective lab-scale separations and direct scale-up from UPLC or HPLC to purification. This column resists mechanical chromatographic bed failure.
  • Usable across a mobile phase pH range of 1 to 12.
  • Trifunctionally bonded C18 ligand for universal HPLC separations.
  • Superior low pH stability and ultra-low column bleed.
  • Improved output and recovery.
  • Faster, more effective lab-scale separations.
  • Direct scale-up from UPLC or HPLC to lab-scale purification.
  • Resistant to mechanical chromatographic bed failure.
  • Consistent, predictable density profiles and cavity avoidance.
Catalog No. 50-782-053 Supplier Waters Corp Supplier No. 186003894
The XBridge BEH C18 sorbent facilitates HPLC method development, usable across a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand provides universal HPLC separations with superior low pH stability and ultra-low column bleed. The Optimum Bed Density (OBD) design enables faster, more effective lab-scale separations and direct scale-up from UPLC or HPLC to purification. This column resists mechanical chromatographic bed failure.
  • Usable across a mobile phase pH range of 1 to 12.
  • Trifunctionally bonded C18 ligand for universal HPLC separations.
  • Superior low pH stability and ultra-low column bleed.
  • Improved output and recovery.
  • Faster, more effective lab-scale separations.
  • Direct scale-up from UPLC or HPLC to lab-scale purification.
  • Resistant to mechanical chromatographic bed failure.
  • Consistent, predictable density profiles and cavity avoidance.