Learn More
Waters Corp Waters Corporation XBridge Prep Column, C18 BEH, 10 μm, 19 x 50 mm
Catalog No. 50782052 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 sorbent is a versatile tool for HPLC method development, functional across a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand offers a universal column suitable for most HPLC separations.
  • Workhorse for HPLC method development, usable over a mobile phase pH range of 1 to 12.
  • Universal column for most HPLC separations due to trifunctionally bonded C18 ligand.
  • Widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed.
Catalog No. 50-782-052 Supplier Waters Corp Supplier No. 186003893
The XBridge BEH C18 sorbent is a versatile tool for HPLC method development, functional across a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand offers a universal column suitable for most HPLC separations.
  • Workhorse for HPLC method development, usable over a mobile phase pH range of 1 to 12.
  • Universal column for most HPLC separations due to trifunctionally bonded C18 ligand.
  • Widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed.