Learn More
Waters Corp XBridge BEH C18 OBD Prep Column, 130Å, 10 µm, 50 mm X 100 mm, 1/pk
Catalog No. 50782061 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1-12). The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed.
Catalog No. 50-782-061 Supplier Waters Corp Supplier No. 186003902
Add to Cart
Add to Cart
Request a Quote
As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1-12). The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed.