Learn More
Waters Corp Prep Guard Cartridge, C18, 10 μm, 19 mm x 10 mm
Catalog No. 50782051 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard cartridge features C18 sorbent, ideal for HPLC method development across a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand provides a universal column for most HPLC separations, offering a wide usable pH range, superior low pH stability, and ultra-low column bleed.
  • Functions as a workhorse for HPLC method development.
  • Suitable for use across a mobile phase pH range of 1 to 12.
  • Features a trifunctionally bonded C18 ligand for universal HPLC separations.
  • Offers wide usable pH range, superior low pH stability, and ultra-low column bleed.
  • Improves column longevity by acting as protective lab equipment.
  • Installed between the injector and the prep column to prevent contaminants.
  • Manufactured with high-performance packing materials.
  • Recommended for biological fluid cleanup and/or temperature control.
Catalog No. 50-782-051 Supplier Waters Corp Supplier No. 186003892
This guard cartridge features C18 sorbent, ideal for HPLC method development across a mobile phase pH range of 1 to 12. Its trifunctionally bonded C18 ligand provides a universal column for most HPLC separations, offering a wide usable pH range, superior low pH stability, and ultra-low column bleed.
  • Functions as a workhorse for HPLC method development.
  • Suitable for use across a mobile phase pH range of 1 to 12.
  • Features a trifunctionally bonded C18 ligand for universal HPLC separations.
  • Offers wide usable pH range, superior low pH stability, and ultra-low column bleed.
  • Improves column longevity by acting as protective lab equipment.
  • Installed between the injector and the prep column to prevent contaminants.
  • Manufactured with high-performance packing materials.
  • Recommended for biological fluid cleanup and/or temperature control.