Learn More
Waters Corp Waters OBD Prep Column, C18, 10 μm, 50 mm × 150 mm
Catalog No. 50782058 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This BEH C18 sorbent column is designed for HPLC method development, compatible with mobile phase pH 1-12. It offers universal applicability for most HPLC separations, featuring superior low pH stability, minimal column bleed, and exceptional mechanical resistance. It enhances productivity in high-resolution, high-speed screening for preparative HPLC.
  • Compatible with mobile phase pH 1-12.
  • Trifunctionally bonded C18 ligand for universal HPLC separations.
  • Superior low pH stability and ultra-low column bleed.
  • Exceptional resistance to mechanical bed failure, ensuring consistent performance and extended lifetime.
  • Maximizes productivity in preparative HPLC through high-resolution, high-speed screening, and direct scalability.
Catalog No. 50-782-058 Supplier Waters Corp Supplier No. 186003899
Add to Cart
Add to Cart
Request a Quote
This BEH C18 sorbent column is designed for HPLC method development, compatible with mobile phase pH 1-12. It offers universal applicability for most HPLC separations, featuring superior low pH stability, minimal column bleed, and exceptional mechanical resistance. It enhances productivity in high-resolution, high-speed screening for preparative HPLC.
  • Compatible with mobile phase pH 1-12.
  • Trifunctionally bonded C18 ligand for universal HPLC separations.
  • Superior low pH stability and ultra-low column bleed.
  • Exceptional resistance to mechanical bed failure, ensuring consistent performance and extended lifetime.
  • Maximizes productivity in preparative HPLC through high-resolution, high-speed screening, and direct scalability.