Learn More
Waters Corp Waters Prep Column, C18, 5 μm, 19 mm × 150 mm, 130 Å
Catalog No. 50781435 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 sorbent is ideal for HPLC method development, offering a mobile phase pH range of 1–12. Its trifunctionally bonded C18 ligand ensures a universal column for most HPLC separations, delivering superior low pH stability and ultra-low column bleed. Optimum Bed Density (OBD) technology enables fast, efficient lab-scale separations, increasing throughput and allowing direct scaling from UPLC, UHPLC, or HPLC screening to purification.
  • Trifunctionally bonded C18 ligand for universal HPLC separations.
  • Operates within a wide pH range of 1–12.
  • Ensures superior low pH stability and ultra-low column bleed.
  • Optimum Bed Density (OBD) technology for extended column lifetimes and consistent performance.
  • Facilitates direct scaling from analytical to lab-scale purification.
  • Access to a preparative columns calculator for scale-up assistance.
Catalog No. 50-781-435 Supplier Waters Corp Supplier No. 186002979
The XBridge BEH C18 sorbent is ideal for HPLC method development, offering a mobile phase pH range of 1–12. Its trifunctionally bonded C18 ligand ensures a universal column for most HPLC separations, delivering superior low pH stability and ultra-low column bleed. Optimum Bed Density (OBD) technology enables fast, efficient lab-scale separations, increasing throughput and allowing direct scaling from UPLC, UHPLC, or HPLC screening to purification.
  • Trifunctionally bonded C18 ligand for universal HPLC separations.
  • Operates within a wide pH range of 1–12.
  • Ensures superior low pH stability and ultra-low column bleed.
  • Optimum Bed Density (OBD) technology for extended column lifetimes and consistent performance.
  • Facilitates direct scaling from analytical to lab-scale purification.
  • Access to a preparative columns calculator for scale-up assistance.