Learn More
Waters Corp Waters Prep Column, BEH C18, 5 μm, 19 x 100 mm
Catalog No. 50781434 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This preparative column is designed for efficient, lab-scale separations and robust purification. It features BEH Technology™, known for excellent ruggedness and longevity across a wide pH range. It is ideal for scaling up methods from analytical to preparative High-Performance Liquid Chromatography (HPLC) while maintaining chromatographic equivalence.
  • Optimum bed density (OBD) design ensures predictable, uniform density profiles and resistance to mechanical bed failure.
  • Increased productivity for fast, efficient, lab-scale separations.
  • Direct scalability from UPLC, UHPLC, or HPLC to lab-scale purification, ensuring chromatographic equivalence.
  • Robust BEH Technology™ provides excellent ruggedness and longevity across a wide pH range.
  • Delivers consistent column-to-column performance, reducing overall costs.
Catalog No. 50-781-434 Supplier Waters Corp Supplier No. 186002978
This preparative column is designed for efficient, lab-scale separations and robust purification. It features BEH Technology™, known for excellent ruggedness and longevity across a wide pH range. It is ideal for scaling up methods from analytical to preparative High-Performance Liquid Chromatography (HPLC) while maintaining chromatographic equivalence.
  • Optimum bed density (OBD) design ensures predictable, uniform density profiles and resistance to mechanical bed failure.
  • Increased productivity for fast, efficient, lab-scale separations.
  • Direct scalability from UPLC, UHPLC, or HPLC to lab-scale purification, ensuring chromatographic equivalence.
  • Robust BEH Technology™ provides excellent ruggedness and longevity across a wide pH range.
  • Delivers consistent column-to-column performance, reducing overall costs.