Learn More
Waters Corp Waters OBD Prep Column, BEH C18, 130Å, 5 μm, 30 mm x 150 mm
Catalog No. 50781651 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C18 sorbent is ideal for HPLC method development, compatible with mobile phase pH from 1 to 12. Its trifunctionally bonded C18 ligand provides a universal column for various HPLC separations, ensuring a wide usable pH range, excellent low pH stability, and minimal column bleed. This preparative column extends analytical column lifespan by removing impurities, maintaining high system separation efficiency and performance, and offers direct scalability from analytical to preparative chromatography.
  • Usable with mobile phase pH from 1 to 12.
  • Provides superior low pH stability and ultra-low column bleed.
  • Increases longevity of analytical columns by eliminating impurities.
  • Ensures system separation efficiency and improves analytical performance.
  • Offers uncompromised scale-up from analytical to preparative applications.
Catalog No. 50-781-651 Supplier Waters Corp Supplier No. 186003284
Add to Cart
Add to Cart
Request a Quote
The XBridge BEH C18 sorbent is ideal for HPLC method development, compatible with mobile phase pH from 1 to 12. Its trifunctionally bonded C18 ligand provides a universal column for various HPLC separations, ensuring a wide usable pH range, excellent low pH stability, and minimal column bleed. This preparative column extends analytical column lifespan by removing impurities, maintaining high system separation efficiency and performance, and offers direct scalability from analytical to preparative chromatography.
  • Usable with mobile phase pH from 1 to 12.
  • Provides superior low pH stability and ultra-low column bleed.
  • Increases longevity of analytical columns by eliminating impurities.
  • Ensures system separation efficiency and improves analytical performance.
  • Offers uncompromised scale-up from analytical to preparative applications.