Learn More
Waters Corp Waters XBridge BEH C18 OBD Prep Column, 130Å, 5 μm, 19 mm X 250 mm
Catalog No. 50782138 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are designed for ruggedness and longevity in LC methods, allowing chromatographers to use mobile phases in a wide pH range. Built with Optimum Bed Density (OBD) technology, they boost productivity with higher recoveries and longer column lifetimes, enabling fast, efficient, lab-scale separations and direct scalability from UPLC, HPLC, and UHPLC screening to lab-scale purification.
  • C18 sorbent for wide mobile phase pH (1 to 12).
  • Trifunctionally bonded C18 ligand for universal HPLC separations.
  • Superior low pH stability and ultra-low column bleed.
  • High pH and temperature stability for increased basic compound loading.
  • OBD technology provides higher recoveries, extended column lifetimes.
  • Direct scalability from UPLC, HPLC, and UHPLC to lab-scale purification.
  • Resists mechanical bed failure, ensures consistent performance and reduced costs.
Catalog No. 50-782-138 Supplier Waters Corp Supplier No. 186004021
Add to Cart
Add to Cart
Request a Quote
These columns are designed for ruggedness and longevity in LC methods, allowing chromatographers to use mobile phases in a wide pH range. Built with Optimum Bed Density (OBD) technology, they boost productivity with higher recoveries and longer column lifetimes, enabling fast, efficient, lab-scale separations and direct scalability from UPLC, HPLC, and UHPLC screening to lab-scale purification.
  • C18 sorbent for wide mobile phase pH (1 to 12).
  • Trifunctionally bonded C18 ligand for universal HPLC separations.
  • Superior low pH stability and ultra-low column bleed.
  • High pH and temperature stability for increased basic compound loading.
  • OBD technology provides higher recoveries, extended column lifetimes.
  • Direct scalability from UPLC, HPLC, and UHPLC to lab-scale purification.
  • Resists mechanical bed failure, ensures consistent performance and reduced costs.