Learn More
Waters Corp Waters Preparative Column, C18, 5 μm, 130Å, 30 × 50 mm
Supplier:  Waters Corp 186002980
Encompass_Preferred
The XBridge BEH C18 sorbent is a workhorse for HPLC method development, usable across a mobile phase pH range of 1 to 12. It offers a universal column for most HPLC separations, providing wide pH range, superior low pH stability, and ultra-low column bleed. Optimum Bed Density (OBD) preparative columns increase efficiency, enabling immediate scaling from UPLC or HPLC screening to lab-scale purification.
  • Universal C18 ligand for HPLC separations.
  • Widest usable pH range (1 to 12) and superior low pH stability.
  • OBD design increases efficiency and stability.
  • Resistant to mechanical chromatographic bed failure.
Catalog No. 50-781-436