Learn More
Waters Corp Guard Cartridge, XBridge, C8, 5 μm, 130 Å
Supplier:  Waters Corp 186002992
Encompass_Preferred
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. It is preferred for method development due to the speed offered by a less retentive column and improved peak shape and chromatographic performance. The trifunctionally bonded sorbent provides the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • Less retentive than BEH C18 sorbent.
  • Improves peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent.
  • Widest usable pH range (1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed.
Catalog No. 50-781-448