Learn More
Waters Corp Waters XBridge Preparative Column, BEH C8, 19 x 100 mm, 10 μm
Catalog No. 50782129 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XBridge Preparative Column features BEH C8 chemistry, dimensions of 19 x 100 mm, and a 10 μm particle size. Designed with OBD Technology, it ensures uniform, optimally packed beds throughout the column’s length and diameter for preparative chromatography.
  • Ensures uniform, optimally packed column beds.
  • Direct scalability from analytical to preparative chromatography.
  • Robust column longevity.
  • Consistent column-to-column performance.
  • Good resolution and peak shape.
  • High mass loading for unparalleled preparative chromatography results.
  • Uncompromised scale-up from UPLC to preparative.
  • Maximizes recoveries for isolation of targeted small molecules and biomolecules.
  • Reduces risk of additional preparative method development.
Catalog No. 50-782-129 Supplier Waters Corp Supplier No. 186004008
This XBridge Preparative Column features BEH C8 chemistry, dimensions of 19 x 100 mm, and a 10 μm particle size. Designed with OBD Technology, it ensures uniform, optimally packed beds throughout the column’s length and diameter for preparative chromatography.
  • Ensures uniform, optimally packed column beds.
  • Direct scalability from analytical to preparative chromatography.
  • Robust column longevity.
  • Consistent column-to-column performance.
  • Good resolution and peak shape.
  • High mass loading for unparalleled preparative chromatography results.
  • Uncompromised scale-up from UPLC to preparative.
  • Maximizes recoveries for isolation of targeted small molecules and biomolecules.
  • Reduces risk of additional preparative method development.