Learn More
Waters Corp Waters Corporation XBridge Preparative Column, BEH C8 Chemistry, 10 μm Particle Size, 19 x 150 mm
Catalog No. 50782130 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Preparative Column is an OBD (Optimum Bed Density) column designed for preparative chromatography applications. It features BEH C8 chemistry and a 10 μm particle size, providing reliable performance for separation.
  • Designed for preparative chromatography applications.
  • Features BEH C8 chemistry.
  • Offers a 10 μm particle size.
  • Dimensions are 19 x 150 mm.
  • Utilizes OBD column technology for optimal performance.
Catalog No. 50-782-130 Supplier Waters Corp Supplier No. 186004009
The XBridge Preparative Column is an OBD (Optimum Bed Density) column designed for preparative chromatography applications. It features BEH C8 chemistry and a 10 μm particle size, providing reliable performance for separation.
  • Designed for preparative chromatography applications.
  • Features BEH C8 chemistry.
  • Offers a 10 μm particle size.
  • Dimensions are 19 x 150 mm.
  • Utilizes OBD column technology for optimal performance.