Learn More
Waters Corp Waters XBridge Preparative Column, BEH C8, 10 μm, 19 x 50 mm, OBD
Supplier:  Waters Corp 186004007
Encompass_Preferred
Waters™ OBD Preparative Columns bridge the performance gap in preparative liquid chromatography (Prep LC) with uniform, optimal packed bed density. This technology ensures direct scalability from analytical to preparative separations, excellent column lifetime, and consistent performance, leading to good resolution, peak shape, and mass loading for unparalleled preparative chromatography results.
  • Ensures uniform, optimal packed bed density.
  • Allows direct scalability from analytical to preparative separations.
  • Offers excellent column lifetime.
  • Provides consistent column-to-column performance.
  • Delivers good resolution, peak shape, and mass loading.
  • Aims for on-target results for successful Prep LC purification.
Catalog No. 50-782-128