Learn More
Waters Corp XBridge OBD Preparative Column, BEH C8, 10 μm, 50 x 250 mm
Catalog No. 50782137 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XBridge OBD Preparative Column utilizes BEH (Ethylene-Bridged Hybrid) particles, with a C8 chemistry providing less retentivity, suitable for the retention of strongly hydrophobic compounds. It has a particle size of 10 μm and dimensions of 50 x 250 mm.
  • Uniform, optimally packed column beds deliver ideal outcomes in Prep LC.
  • Direct scalability from analytical to Preparative LC.
  • Excellent column lifetime ensures long-term performance.
  • Consistent column-to-column performance leads to good resolution, peak shape, and mass loading.
  • Trust in on-target results for successful Prep LC purification with OBD Technology.
  • Maximize recoveries with robust packings designed for isolation of targeted small molecules and biomolecules.
  • Decrease risk of additional Prep method development.
Catalog No. 50-782-137 Supplier Waters Corp Supplier No. 186004016
Add to Cart
Add to Cart
Request a Quote
This XBridge OBD Preparative Column utilizes BEH (Ethylene-Bridged Hybrid) particles, with a C8 chemistry providing less retentivity, suitable for the retention of strongly hydrophobic compounds. It has a particle size of 10 μm and dimensions of 50 x 250 mm.
  • Uniform, optimally packed column beds deliver ideal outcomes in Prep LC.
  • Direct scalability from analytical to Preparative LC.
  • Excellent column lifetime ensures long-term performance.
  • Consistent column-to-column performance leads to good resolution, peak shape, and mass loading.
  • Trust in on-target results for successful Prep LC purification with OBD Technology.
  • Maximize recoveries with robust packings designed for isolation of targeted small molecules and biomolecules.
  • Decrease risk of additional Prep method development.