Learn More
Waters Corp Waters XBridge Preparative Column, BEH C8, 50 × 50 mm, 10 μm
Catalog No. 50782134 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is an XBridge Preparative OBD Column with BEH C8 chemistry, designed for preparative chromatography. It has dimensions of 50 × 50 mm and a particle size of 10 μm.
  • OBD column type
  • BEH C8 chemistry
  • Dimensions: 50 × 50 mm
  • Particle size: 10 μm
Catalog No. 50-782-134 Supplier Waters Corp Supplier No. 186004013
Add to Cart
Add to Cart
Request a Quote
This is an XBridge Preparative OBD Column with BEH C8 chemistry, designed for preparative chromatography. It has dimensions of 50 × 50 mm and a particle size of 10 μm.
  • OBD column type
  • BEH C8 chemistry
  • Dimensions: 50 × 50 mm
  • Particle size: 10 μm