Learn More
Waters Corp Waters XBridge BEH C8 Prep Column, 130Å, 10 μm, 30 mm x 150 mm
Catalog No. 50782132 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C8 Prep Column features a less retentive C8 sorbent compared to C18, making it ideal for method development by offering increased speed while enhancing peak shape and chromatographic performance. Its trifunctionally bonded sorbent provides a wide usable pH range of 1 to 12, ensuring superior low pH stability and ultra-low column bleed for reliable results.
  • Less retentive C8 sorbent, ideal for faster method development.
  • Improves peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent with wide pH range (1 to 12).
  • Superior low pH stability and ultra-low column bleed.
  • Hybrid particle matrix with spherical shape.
  • Particle size: 10 μm.
  • Pore size: 130 Å.
  • Dimensions: 30 mm inner diameter, 150 mm length.
  • Suitable for HPLC preparative, LC/MS, and LC techniques.
Catalog No. 50-782-132 Supplier Waters Corp Supplier No. 186004011
Add to Cart
Add to Cart
Request a Quote
The XBridge BEH C8 Prep Column features a less retentive C8 sorbent compared to C18, making it ideal for method development by offering increased speed while enhancing peak shape and chromatographic performance. Its trifunctionally bonded sorbent provides a wide usable pH range of 1 to 12, ensuring superior low pH stability and ultra-low column bleed for reliable results.
  • Less retentive C8 sorbent, ideal for faster method development.
  • Improves peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent with wide pH range (1 to 12).
  • Superior low pH stability and ultra-low column bleed.
  • Hybrid particle matrix with spherical shape.
  • Particle size: 10 μm.
  • Pore size: 130 Å.
  • Dimensions: 30 mm inner diameter, 150 mm length.
  • Suitable for HPLC preparative, LC/MS, and LC techniques.