Learn More
Waters Corp Waters XBridge OBD Prep Column, 10 μm, 130 Å, 50 mm × 100 mm
Catalog No. 50782135 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. It is designed to offer speed during method development while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent provides the widest usable pH range (1–12), superior low pH stability, and ultra–low column bleed.
  • Less retentive compared to BEH C18 sorbent, offering speed during method development.
  • Improves peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent.
  • Widest usable pH range (1–12).
  • Superior low pH stability.
  • Ultra–low column bleed.
Catalog No. 50-782-135 Supplier Waters Corp Supplier No. 186004014
Add to Cart
Add to Cart
Request a Quote
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. It is designed to offer speed during method development while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent provides the widest usable pH range (1–12), superior low pH stability, and ultra–low column bleed.
  • Less retentive compared to BEH C18 sorbent, offering speed during method development.
  • Improves peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent.
  • Widest usable pH range (1–12).
  • Superior low pH stability.
  • Ultra–low column bleed.