Learn More
Waters Corp Waters OBD Prep Column, C8, 10 μm, 19 mm × 250 mm, 130 Å
Catalog No. 50782131 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This OBD Prep Column utilizes a C8 sorbent, designed for faster method development and enhanced chromatographic performance. Its trifunctionally bonded sorbent provides a wide usable pH range, excellent low pH stability, and minimal column bleed.
  • Enables faster method development.
  • Improves peak shape and chromatographic performance.
  • Features a trifunctionally bonded sorbent.
  • Provides a wide usable pH range of 1 to 12.
  • Offers superior stability at low pH.
  • Ensures ultra-low column bleed.
Catalog No. 50-782-131 Supplier Waters Corp Supplier No. 186004010
Add to Cart
Add to Cart
Request a Quote
This OBD Prep Column utilizes a C8 sorbent, designed for faster method development and enhanced chromatographic performance. Its trifunctionally bonded sorbent provides a wide usable pH range, excellent low pH stability, and minimal column bleed.
  • Enables faster method development.
  • Improves peak shape and chromatographic performance.
  • Features a trifunctionally bonded sorbent.
  • Provides a wide usable pH range of 1 to 12.
  • Offers superior stability at low pH.
  • Ensures ultra-low column bleed.