Learn More
Waters Corp Waters Corporation XBridge Preparative Column, BEH C8, 130Å, 5 μm, 50 mm × 100 mm
Catalog No. 50782073 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XBridge Preparative Column with BEH C8 chemistry is designed for high-performance preparative liquid chromatography, offering uniform packed bed density for reliable and efficient purification.
  • Uniform, optimum packed bed density ensures reliable and efficient purification.
  • Direct scalability from analytical to preparative methods.
  • Exceptional column lifetime and consistent performance.
  • Suitable for a wide variety of compounds.
  • Delivers superior peak shape for basic analytes at elevated pH.
  • Stable across a wide pH range and at high temperatures.
  • Robust packing for isolation of targeted small molecules and biomolecules.
  • Maximizes recoveries and decreases risk in method development.
  • Provides uncompromised scale-up from UPLC™ to Prep.
Catalog No. 50-782-073 Supplier Waters Corp Supplier No. 186003938
Add to Cart
Add to Cart
Request a Quote
This XBridge Preparative Column with BEH C8 chemistry is designed for high-performance preparative liquid chromatography, offering uniform packed bed density for reliable and efficient purification.
  • Uniform, optimum packed bed density ensures reliable and efficient purification.
  • Direct scalability from analytical to preparative methods.
  • Exceptional column lifetime and consistent performance.
  • Suitable for a wide variety of compounds.
  • Delivers superior peak shape for basic analytes at elevated pH.
  • Stable across a wide pH range and at high temperatures.
  • Robust packing for isolation of targeted small molecules and biomolecules.
  • Maximizes recoveries and decreases risk in method development.
  • Provides uncompromised scale-up from UPLC™ to Prep.