Learn More
Waters Corp Waters XBridge BEH C8 OBD Prep Column, 130Å, 5 μm, 30 mm × 150 mm
Catalog No. 50781535 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This C8 sorbent is less retentive compared to the C18 sorbent. It offers speed for method development, while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent provides the widest usable pH range (1–12), superior low pH stability, and ultra-low column bleed.
  • Less retentive C8 sorbent for faster method development.
  • Improves peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent provides the widest usable pH range (1–12).
  • Superior low pH stability.
  • Ultra-low column bleed.
  • Good universal column choice for a wide variety of compounds.
  • Provides exceptional peak shape for basic analytes at elevated pH.
  • Stable across a wide pH range and at high temperatures.
  • Suitable for the retention of strongly hydrophobic compounds.
Catalog No. 50-781-535 Supplier Waters Corp Supplier No. 186003083
Add to Cart
Add to Cart
Request a Quote
This C8 sorbent is less retentive compared to the C18 sorbent. It offers speed for method development, while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent provides the widest usable pH range (1–12), superior low pH stability, and ultra-low column bleed.
  • Less retentive C8 sorbent for faster method development.
  • Improves peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent provides the widest usable pH range (1–12).
  • Superior low pH stability.
  • Ultra-low column bleed.
  • Good universal column choice for a wide variety of compounds.
  • Provides exceptional peak shape for basic analytes at elevated pH.
  • Stable across a wide pH range and at high temperatures.
  • Suitable for the retention of strongly hydrophobic compounds.