Learn More
Waters Corp XBridge BEH HILIC OBD Prep Column, 130Å, 5 µm, 19 mm X 250 mm, 1/pk
Supplier:  Waters Corp 186004730
Encompass_Preferred
XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions. More rugged than silica based HILIC phases, the BEH particle’s chemical stability results in long column lifetimes.
Catalog No. 50-466-054
Add to Cart
Request a Quote