Learn More
Waters Corp Waters XBridge Preparative Column, BEH HILIC, 30 x 50 mm, 5 μm
Catalog No. 50466051 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Preparative Column, BEH HILIC, is designed for high-performance liquid chromatography. It provides excellent retention for very polar, basic, and water-soluble analytes. This column is specifically developed and tested for HILIC separations using mobile phases containing a high concentration of organic solvent.
  • Excellent retention for very polar, basic, water soluble analytes.
  • Designed and tested for HILIC separations using mobile phases containing high concentration of organic solvent.
Catalog No. 50-466-051 Supplier Waters Corp Supplier No. 186004727
The XBridge Preparative Column, BEH HILIC, is designed for high-performance liquid chromatography. It provides excellent retention for very polar, basic, and water-soluble analytes. This column is specifically developed and tested for HILIC separations using mobile phases containing a high concentration of organic solvent.
  • Excellent retention for very polar, basic, water soluble analytes.
  • Designed and tested for HILIC separations using mobile phases containing high concentration of organic solvent.