Learn More
Waters Corp Waters HILIC OBD Prep Column, BEH, 130Å, 5 μm, 19 mm X 50 mm
Supplier:  Waters Corp 186004724
Encompass_Preferred
XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions. More rugged than silica based HILIC phases, the BEH particle's chemical stability results in long column lifetimes.
  • Contains unbonded BEH substrate.
  • Designed to retain and separate very polar compounds.
  • Optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions.
  • More rugged than silica-based HILIC phases.
  • Particle's chemical stability results in long column lifetimes.
Catalog No. 50-466-048