Learn More
Waters Corp Preparative Column, BEH HILIC, 5 μm, 30 mm x 150 mm, 130 Å
Supplier:  Waters Corp 186004729
Encompass_Preferred
This preparative column features unbonded BEH substrate, designed for retaining and separating very polar compounds. It is optimized for efficient, reproducible separations under HPLC HILIC conditions, offering long column lifetimes due to the BEH particle's chemical stability.
  • Excellent retention for very polar, basic, water soluble analytes.
  • Optimized for HILIC separations with high organic solvent mobile phases.
  • Universal column choice for a wide variety of compounds.
  • Exceptional peak shape for basic analytes at elevated pH.
  • Stable across a wide pH range and at high temperatures.
  • Uniform, optimum packed bed density throughout the column.
  • Direct scalability from analytical to preparative, with excellent lifetime and consistent performance.
  • Good resolution, peak shape, and mass loading for unparalleled preparative chromatography.
Catalog No. 50-466-053
Add to Cart
Request a Quote