Learn More
Waters Corp WATERS CORPORATION Prep Guard Cartridge, 5 μm, 130 Å, 19 mm × 10 mm
Catalog No. 50781707 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Phenyl columns offer complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. This product features the most chemically stable commercially available Phenyl sorbent, providing ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Offers complementary selectivity for polyaromatic compounds.
  • Chemically stable phenyl sorbent for ultra-low column bleed and long lifetimes.
  • Ensures excellent peak shape.
  • Phenyl chemistry with reversed phase separation.
  • Hybrid particle substrate, pH range 1 to 12.
  • Temperature limits: 80°C (low pH) / 60°C (high pH).
  • Maximum pressure: 6000 psi (415 Bar).
  • Particle size: 5 μm; Pore size: 130 Å.
  • Prep guard cartridge for HPLC preparative systems.
Catalog No. 50-781-707 Supplier Waters Corp Supplier No. 186003355
XBridge BEH Phenyl columns offer complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. This product features the most chemically stable commercially available Phenyl sorbent, providing ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Offers complementary selectivity for polyaromatic compounds.
  • Chemically stable phenyl sorbent for ultra-low column bleed and long lifetimes.
  • Ensures excellent peak shape.
  • Phenyl chemistry with reversed phase separation.
  • Hybrid particle substrate, pH range 1 to 12.
  • Temperature limits: 80°C (low pH) / 60°C (high pH).
  • Maximum pressure: 6000 psi (415 Bar).
  • Particle size: 5 μm; Pore size: 130 Å.
  • Prep guard cartridge for HPLC preparative systems.