Learn More
Waters Corp Waters XBridge BEH Phenyl Prep Guard Cartridge, 130Å, 5 μm, 10 mm X 10 mm
Catalog No. 50781706 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Prep Guard Cartridge offers complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
  • Provides complementary selectivity to C18 stationary phases
  • Optimized for polyaromatic compounds
  • Features a chemically stable phenyl sorbent
  • Ensures ultra-low column bleed
  • Offers long column lifetimes
  • Delivers excellent peak shape
Catalog No. 50-781-706 Supplier Waters Corp Supplier No. 186003354
This Prep Guard Cartridge offers complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
  • Provides complementary selectivity to C18 stationary phases
  • Optimized for polyaromatic compounds
  • Features a chemically stable phenyl sorbent
  • Ensures ultra-low column bleed
  • Offers long column lifetimes
  • Delivers excellent peak shape