Learn More
Waters Corp Waters Prep Column, Phenyl, 5 μm, 50 mm × 50 mm
Catalog No. 50782071 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This preparative column offers complementary selectivity to C18 stationary phases for polyaromatic compounds. It uses a stable Phenyl sorbent for ultra-low column bleed, extended lifetimes, and excellent peak shapes. Constructed with hybrid particle substrate and phenyl-hexyl bonding, it has a spherical particle with low silanol activity. It operates within a pH range of 1 to 12 and handles temperatures up to 80°C (low pH) or 60°C (high pH), with a maximum pressure of 6000 psi (415 Bar). Suitable for HPLC preparative and LC/MS techniques.
  • Complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
  • Most chemically stable commercially available Phenyl sorbent on the market.
  • Provides ultra-low column bleed.
  • Offers long column lifetimes.
  • Ensures excellent peak shape.
  • Endcapped.
  • Low silanol activity.
  • Spherical particle shape.
Catalog No. 50-782-071 Supplier Waters Corp Supplier No. 186003936
Add to Cart
Add to Cart
Request a Quote
This preparative column offers complementary selectivity to C18 stationary phases for polyaromatic compounds. It uses a stable Phenyl sorbent for ultra-low column bleed, extended lifetimes, and excellent peak shapes. Constructed with hybrid particle substrate and phenyl-hexyl bonding, it has a spherical particle with low silanol activity. It operates within a pH range of 1 to 12 and handles temperatures up to 80°C (low pH) or 60°C (high pH), with a maximum pressure of 6000 psi (415 Bar). Suitable for HPLC preparative and LC/MS techniques.
  • Complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.
  • Most chemically stable commercially available Phenyl sorbent on the market.
  • Provides ultra-low column bleed.
  • Offers long column lifetimes.
  • Ensures excellent peak shape.
  • Endcapped.
  • Low silanol activity.
  • Spherical particle shape.